siRNA Details
| Gene Symbol: GUSBP11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BX538181.1 | - | 12 | 3028 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000421064.5 | - | 12 | 3031 | Chr. 22 - 23653169 - 23717347 [-] on Build GRCh38 | |

GUSBP11
Genomic Map