siRNA Details
| Gene Symbol: CDC37L1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121597.1 | 2 | 275 | |||
| NR_121598.1 | 2 | 197 | ||||
| GenBank mRNA | CK820219.1 | - | 2 | 188 | ||
| CR737367.1 | - | 2 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC37L1-AS1| Gene Symbol: CDC37L1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121597.1 | 2 | 275 | |||
| NR_121598.1 | 2 | 197 | ||||
| GenBank mRNA | CK820219.1 | - | 2 | 188 | ||
| CR737367.1 | - | 2 | 274 | |||