siRNA Details
| Gene Symbol: LOC101929607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110039.1 | 3 | 820 | |||
| GenBank mRNA | BC042024.1 | - | 2 | 819 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000609293.1 | - | 3 | 820 | Chr. 3 - 107430930 - 107463912 [+] on Build GRCh38 | |

LOC101929607
Genomic Map