siRNA Details
| Gene Symbol: FLT3LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK301136.1 | - | 8 | 1297 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000595815.1 | - | 8 | 1297 | Chr. 19 - 49474586 - 49487637 [+] on Build GRCh38 | |

FLT3LG
Genomic Map