siRNA Details
Gene Symbol: ACKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC011588.1 | - | 4 | 952 | ||
BC018716.1 | - | 4 | 952 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000471537.3 | - | 4 | 952 | Chr. 3 - 42809472 - 42900336 [+] on Build GRCh38 |
