siRNA Details
| Gene Symbol: CDKL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK294559.1 | - | 10 | 1436 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000520515.5 | - | 10 | 1436 | Chr. 5 - 134205614 - 134371043 [-] on Build GRCh38 | |

CDKL3
Genomic Map