siRNA Details
| Gene Symbol: FLJ12120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK022182.1 | - | 1 | 23 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000615128.1 | - | 926 | Chr. - - [-] on Build | ||

FLJ12120
Genomic Map