siRNA Details
| Gene Symbol: LINC00547 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040244.1 | 1, 2 | 34 | |||
| GenBank mRNA | AK092066.1 | - | 1, 2 | 34 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000612438.1 | - | 34 | Chr. - - [-] on Build | ||

LINC00547
Genomic Map