siRNA Details
| Gene Symbol: C9orf173-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038389.1 | 4 | 1125 | |||
| GenBank mRNA | AK097764.1 | - | 4 | 1125 | ||
| Gene Symbol: FAM166A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006717108.2 | XP_006717171.2 | 1 | 399 | ||
| XM_011518694.1 | XP_011516996.1 | 1 | 399 | |||
| XM_011518695.1 | XP_011516997.1 | 1 | 399 | |||
| XM_011518696.2 | XP_011516998.1 | 1 | 399 | |||
| XM_011518697.1 | XP_011516999.1 | 1 | 399 | |||
| XM_017014718.1 | XP_016870207.1 | 1 | 399 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000612170.1 | - | 1125 | Chr. - - [-] on Build | ||

C9orf173-AS1
Genomic Map