siRNA Details
| Gene Symbol: LOC100505942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104656.1 | 2 | 823 | |||
| GenBank mRNA | BG210561.1 | - | 1 | 399 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000621378.1 | - | 753 | Chr. - - [-] on Build | ||

LOC100505942
Genomic Map