siRNA Details
Gene Symbol: LOC400661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122073.1 | 2 | 2188 | |||
GenBank mRNA | AK126829.1 | - | 2 | 2187 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000611889.1 | - | 2188 | Chr. - - [-] on Build |
