siRNA Details
Gene Symbol: LOC101929698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110619.1 | 3 | 1287 | |||
GenBank mRNA | AK097865.1 | - | 3 | 1287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000620523.1 | - | 1287 | Chr. - - [-] on Build |
