siRNA Details
Gene Symbol: LOC101929163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136246.1 | 2 | 465 | |||
GenBank mRNA | BX117162.1 | - | 2 | 354 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000611838.1 | - | 354 | Chr. - - [-] on Build |
