siRNA Details
| Gene Symbol: LOC101926897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120416.1 | 3 | 2281 | |||
| GenBank mRNA | AK124674.1 | - | 3 | 2282 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000615202.1 | - | 2281 | Chr. - - [-] on Build | ||

LOC101926897
Genomic Map