siRNA Details
Gene Symbol: LOC101926897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120416.1 | 3 | 1572 | |||
GenBank mRNA | AK124674.1 | - | 3 | 1573 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000615202.1 | - | 1572 | Chr. - - [-] on Build |
