siRNA Details
| Gene Symbol: TSC22D1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038381.1 | 3 | 904 | |||
| GenBank mRNA | BC036219.1 | - | 1 | 1005 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSC22D1-AS1| Gene Symbol: TSC22D1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038381.1 | 3 | 904 | |||
| GenBank mRNA | BC036219.1 | - | 1 | 1005 | ||