siRNA Details
| Gene Symbol: LOC101929140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120423.1 | 4 | 499 | |||
| GenBank mRNA | AK056182.1 | - | 4 | 488 | ||
| DA634395.1 | - | 4 | 499 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000619407.4 | - | 488 | Chr. - - [-] on Build | ||

LOC101929140
Genomic Map