siRNA Details
Gene Symbol: LOC101929140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120423.1 | 4 | 713 | |||
GenBank mRNA | AK056182.1 | - | 4 | 702 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000619407.4 | - | 702 | Chr. - - [-] on Build |
