siRNA Details
Gene Symbol: LOC101929140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120423.1 | 2 | 199 | |||
GenBank mRNA | AK056182.1 | - | 2 | 188 | ||
DA634395.1 | - | 2 | 199 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101929140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120423.1 | 2 | 199 | |||
GenBank mRNA | AK056182.1 | - | 2 | 188 | ||
DA634395.1 | - | 2 | 199 |