siRNA Details
| Gene Symbol: LOC642929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027472.1 | 1 | 95 | |||
| GenBank mRNA | DQ786189.1 | - | 1 | 95 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000610400.1 | - | 93 | Chr. - - [-] on Build | ||

LOC642929
Genomic Map