siRNA Details
| Gene Symbol: LOC101926905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105056.2 | 2 | 291 | |||
| GenBank mRNA | HY003594.1 | - | 3 | 229 | ||
| HY010779.1 | - | 2 | 284 | |||
| HY018974.1 | - | 3 | 416 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000624778.1 | - | 284 | Chr. - - [-] on Build | ||

LOC101926905
Genomic Map