siRNA Details
Gene Symbol: LOC101926905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_105056.2 | 2 | 324 | |||
GenBank mRNA | HY003594.1 | - | 3 | 262 | ||
HY010779.1 | - | 2 | 317 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000624778.1 | - | 317 | Chr. - - [-] on Build |
