siRNA Details
| Gene Symbol: SCAANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU569004.1 | - | 60 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCAANT1| Gene Symbol: SCAANT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BU569004.1 | - | 60 | |||