siRNA Details
Gene Symbol: LINC00850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109813.1 | 3 | 375 | |||
GenBank mRNA | JX283354.1 | - | 3 | 375 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000629221.1 | - | 375 | Chr. - - [-] on Build |
