siRNA Details
| Gene Symbol: LINC00921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033904.1 | 1 | 1774 | |||
| GenBank mRNA | AK096958.1 | - | 1 | 1740 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000573951.1 | - | 1 | 1799 | Chr. 16 - 3263743 - 3267567 [+] on Build GRCh38 | |

LINC00921
Genomic Map