siRNA Details
| Gene Symbol: PTENP1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103745.2 | 1, 2 | 241 | |||
| GenBank mRNA | BX374997.2 | - | 1, 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PTENP1-AS| Gene Symbol: PTENP1-AS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103745.2 | 1, 2 | 241 | |||
| GenBank mRNA | BX374997.2 | - | 1, 2 | 241 | ||