siRNA Details
| Gene Symbol: SCHLAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104319.1 | 5 | 915 | |||
| NR_104320.1 | 4 | 846 | ||||
| NR_104321.1 | 3 | 579 | ||||
| NR_104322.1 | 7 | 1154 | ||||
| NR_104323.1 | 6 | 1053 | ||||
| NR_104324.1 | 5 | 947 | ||||
| NR_104325.1 | 6 | 1016 | ||||
| GenBank mRNA | JX117418.1 | - | 5 | 915 | ||
| JX117419.1 | - | 4 | 846 | |||
| JX117420.1 | - | 3 | 579 | |||
| JX117421.1 | - | 7 | 1154 | |||
| JX117422.1 | - | 6 | 1053 | |||
| JX117423.1 | - | 5 | 947 | |||
| JX117424.1 | - | 6 | 1016 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000629145.1 | - | 915 | Chr. - - [-] on Build | ||

SCHLAP1
Genomic Map