siRNA Details
| Gene Symbol: 6330403K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ271140.1 | - | 1 | 675 | ||
| AK083923.1 | - | 1 | 682 | |||
| BC069976.1 | - | 1 | 675 | 6518302 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330403K07Rik| Gene Symbol: 6330403K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ271140.1 | - | 1 | 675 | ||
| AK083923.1 | - | 1 | 682 | |||
| BC069976.1 | - | 1 | 675 | 6518302 | ||