siRNA Details
| Gene Symbol: Kcnmb4os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028107.1 | 2 | 704 | |||
| GenBank mRNA | AK006831.1 | - | 2 | 705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnmb4os1| Gene Symbol: Kcnmb4os1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028107.1 | 2 | 704 | |||
| GenBank mRNA | AK006831.1 | - | 2 | 705 | ||