siRNA Details
| Gene Symbol: 4933413J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038005.1 | 8 | 1322 | |||
| GenBank mRNA | AK087762.1 | - | 8 | 1326 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933413J09Rik| Gene Symbol: 4933413J09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038005.1 | 8 | 1322 | |||
| GenBank mRNA | AK087762.1 | - | 8 | 1326 | ||