siRNA Details
| Gene Symbol: 4930562F07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038030.1 | 3 | 646 | |||
| NR_038031.1 | 3 | 574 | ||||
| GenBank mRNA | AK016191.1 | - | 3 | 647 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA466557.1 | - | 3 | 570 | Chr. 1 - 160074822 - 160077977 [-] on Build GRCm38 | |

4930562F07Rik
Genomic Map