siRNA Details
| Gene Symbol: 4921508D12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045802.1 | 1 | 62 | |||
| GenBank mRNA | AK014842.1 | - | 1 | 64 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714363.1 | - | 1 | 64 | Chr. 2 - 132424752 - 132452357 [+] on Build GRCm38 | |

4921508D12Rik
Genomic Map