siRNA Details
Gene Symbol: Crnde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033641.3 | 6 | 999 | |||
NR_110452.1 | 4 | 698 | ||||
GenBank mRNA | AK017076.1 | - | 6 | 999 | ||
CX205694.1 | - | 4 | 409 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crnde | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033641.3 | 6 | 999 | |||
NR_110452.1 | 4 | 698 | ||||
GenBank mRNA | AK017076.1 | - | 6 | 999 | ||
CX205694.1 | - | 4 | 409 |