siRNA Details
| Gene Symbol: 6430628N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018292.1 | - | 2 | 972 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6430628N08Rik| Gene Symbol: 6430628N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018292.1 | - | 2 | 972 | ||