siRNA Details
| Gene Symbol: 1700006J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005678.1 | - | 3 | 496 | ||
| AK011105.1 | - | 4 | 536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700006J14Rik| Gene Symbol: 1700006J14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005678.1 | - | 3 | 496 | ||
| AK011105.1 | - | 4 | 536 | |||