siRNA Details
| Gene Symbol: Fam219aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045726.1 | 1 | 130 | |||
| GenBank mRNA | AK006904.1 | - | 1 | 131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam219aos| Gene Symbol: Fam219aos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045726.1 | 1 | 130 | |||
| GenBank mRNA | AK006904.1 | - | 1 | 131 | ||