siRNA Details
| Gene Symbol: 4930579G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038053.1 | 6 | 866 | |||
| GenBank mRNA | AK016319.1 | - | 6 | 867 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930579G18Rik| Gene Symbol: 4930579G18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038053.1 | 6 | 866 | |||
| GenBank mRNA | AK016319.1 | - | 6 | 867 | ||