siRNA Details
| Gene Symbol: 4930426D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015205.1 | - | 4 | 836 | ||
| AK156636.1 | - | 4 | 867 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930426D05Rik| Gene Symbol: 4930426D05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015205.1 | - | 4 | 836 | ||
| AK156636.1 | - | 4 | 867 | |||