siRNA Details
| Gene Symbol: Plekhd1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037995.1 | 2 | 912 | |||
| GenBank mRNA | AK002366.1 | - | 2 | 912 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhd1os| Gene Symbol: Plekhd1os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_037995.1 | 2 | 912 | |||
| GenBank mRNA | AK002366.1 | - | 2 | 912 | ||