siRNA Details
Gene Symbol: 9530080O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045776.1 | 2 | 704 | |||
NR_045777.1 | 2 | 422 | ||||
GenBank mRNA | AK035647.1 | - | 2 | 704 | ||
CJ060540.1 | - | 2 | 423 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9530080O11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045776.1 | 2 | 704 | |||
NR_045777.1 | 2 | 422 | ||||
GenBank mRNA | AK035647.1 | - | 2 | 704 | ||
CJ060540.1 | - | 2 | 423 |