siRNA Details
| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037053.1 | - | 2 | 65 | ||
| AK158126.1 | - | 1 | 137 | |||
| AK160978.1 | - | 2 | 57 | |||
| BC099946.1 | - | 1 | 42 | 30102988 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9930104L06Rik| Gene Symbol: 9930104L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037053.1 | - | 2 | 65 | ||
| AK158126.1 | - | 1 | 137 | |||
| AK160978.1 | - | 2 | 57 | |||
| BC099946.1 | - | 1 | 42 | 30102988 | ||