siRNA Details
| Gene Symbol: E130304I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033567.1 | 3 | 592 | |||
| GenBank mRNA | AK021412.1 | - | 3 | 594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E130304I02Rik| Gene Symbol: E130304I02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033567.1 | 3 | 592 | |||
| GenBank mRNA | AK021412.1 | - | 3 | 594 | ||