siRNA Details
| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040263.1 | 2 | 550 | |||
| NR_040264.1 | 2 | 546 | ||||
| GenBank mRNA | AK006282.1 | - | 2 | 528 | ||
| AK076883.1 | - | 2 | 545 | |||
| BY706362.1 | - | 2 | 522 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU937321.1 | - | 528 | Chr. - - [-] on Build | ||

Smkr-ps
Genomic Map