siRNA Details
| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040263.1 | 2 | 344 | |||
| NR_040264.1 | 2 | 340 | ||||
| GenBank mRNA | AK006282.1 | - | 2 | 322 | ||
| AK076883.1 | - | 2 | 339 | |||
| BB615595.1 | - | 2 | 336 | |||
| BY706362.1 | - | 2 | 316 | |||

Smkr-ps
Genomic Map