siRNA Details
| Gene Symbol: C230012O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044654.1 | - | 1 | 197 | ||
| AK048707.1 | - | 1 | 666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230012O17Rik| Gene Symbol: C230012O17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044654.1 | - | 1 | 197 | ||
| AK048707.1 | - | 1 | 666 | |||