siRNA Details
| Gene Symbol: 1700031F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI647336.1 | - | 2 | 366 | ||
| AK006579.1 | - | 2 | 327 | |||
| BC099573.1 | - | 2 | 385 | |||
| CN837050.1 | - | 388 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031F10Rik| Gene Symbol: 1700031F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AI647336.1 | - | 2 | 366 | ||
| AK006579.1 | - | 2 | 327 | |||
| BC099573.1 | - | 2 | 385 | |||
| CN837050.1 | - | 388 | ||||