siRNA Details
| Gene Symbol: A630023P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102290.1 | 3 | 501 | |||
| GenBank mRNA | AK041600.1 | - | 4 | 1076 | ||
| AK088579.1 | - | 3 | 502 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630023P12Rik| Gene Symbol: A630023P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102290.1 | 3 | 501 | |||
| GenBank mRNA | AK041600.1 | - | 4 | 1076 | ||
| AK088579.1 | - | 3 | 502 | |||