siRNA Details
| Gene Symbol: 4930542C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005833.1 | - | 393 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930542C16Rik| Gene Symbol: 4930542C16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005833.1 | - | 393 | |||