siRNA Details
Gene Symbol: 4930469G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015528.1 | - | 260 |

More Information
Other Species Targeted


Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930469G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015528.1 | - | 260 |