siRNA Details
| Gene Symbol: 1700104B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018937.1 | - | 2 | 113 | ||
| AV044329.3 | - | 2 | 130 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700104B16Rik| Gene Symbol: 1700104B16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018937.1 | - | 2 | 113 | ||
| AV044329.3 | - | 2 | 130 | |||